Assay Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290148.1 | NP_001277077.1 | 2 - 3 | 266 | 99 | |
| NM_028964.4 | NP_083240.2 | 9 - 10 | 1315 | 99 | ||
| GenBank mRNA | AF399755.1 | - | 2 - 3 | 221 | 99 | |
| AK017102.1 | - | 2 - 3 | 267 | 99 | ||
| AK162238.1 | - | 1 - 2 | 141 | 99 | ||
| BB623950.1 | - | 1 - 2 | 141 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snx29
Genomic Map