Assay Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025992.2 | NP_080268.1 | 2 - 3 | 403 | 148 | |
| GenBank mRNA | AK011047.1 | - | 2 - 3 | 290 | 148 | |
| AK015214.1 | - | 446 | 148 | |||
| AK017888.1 | - | 2 - 3 | 231 | 148 | ||
| AK147960.1 | - | 2 - 3 | 327 | 148 | ||
| AK149648.1 | - | 2 - 3 | 395 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015214.1 transcripts.
Set Membership: |
|

Herc6
Genomic Map