Assay Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025992.2 | NP_080268.1 | 4 - 5 | 705 | 142 | |
| GenBank mRNA | AK011047.1 | - | 4 - 5 | 592 | 142 | |
| AK015214.1 | - | 747 | 141 | |||
| AK017888.1 | - | 4 - 5 | 533 | 142 | ||
| AK149648.1 | - | 4 - 5 | 697 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Herc6
Genomic Map