Assay Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172961.3 | NP_766549.2 | 12 - 13 | 1144 | 59 | |
| GenBank mRNA | AK132282.1 | - | 12 - 13 | 1093 | 59 | |
| AK165348.1 | - | 12 - 13 | 1095 | 59 | ||
| BC058079.1 | - | 12 - 13 | 1084 | 6826741 | 59 | |
| BC058521.1 | - | 12 - 13 | 1084 | 6825487 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abat
Genomic Map