Assay Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172961.3 | NP_766549.2 | 13 - 14 | 1315 | 91 | |
| GenBank mRNA | AK132282.1 | - | 13 - 14 | 1264 | 91 | |
| AK165348.1 | - | 13 - 14 | 1266 | 91 | ||
| BC058079.1 | - | 13 - 14 | 1255 | 6826741 | 91 | |
| BC058521.1 | - | 13 - 14 | 1255 | 6825487 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abat
Genomic Map