Assay Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170978.1 | NP_001164449.1 | 13 - 14 | 1293 | 100 | |
| NM_172961.3 | NP_766549.2 | 14 - 15 | 1461 | 100 | ||
| GenBank mRNA | AK036128.1 | - | 13 - 14 | 1238 | 100 | |
| AK132282.1 | - | 14 - 15 | 1410 | 100 | ||
| AK156736.1 | - | 2 - 3 | 170 | 100 | ||
| AK165348.1 | - | 14 - 15 | 1412 | 100 | ||
| BC058079.1 | - | 14 - 15 | 1401 | 6826741 | 100 | |
| BC058521.1 | - | 14 - 15 | 1401 | 6825487 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abat
Genomic Map