Assay Details
| Gene Symbol: Spen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019763.2 | NP_062737.2 | 11 - 12 | 2278 | 75 | |
| GenBank mRNA | AF156529.1 | - | 10 - 11 | 1780 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spen
Genomic Map