Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400032158.mm.1 |
TC0400003884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032158.mm.1 |
NM_019763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032158.mm.1 |
NM_019763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032160.mm.1 |
TC0400003884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032160.mm.1 |
NM_019763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032160.mm.1 |
NM_019763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032154.mm.1 |
TC0400003884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032154.mm.1 |
NM_019763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032154.mm.1 |
NM_019763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032159.mm.1 |
TC0400003884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032159.mm.1 |
NM_019763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032159.mm.1 |
NM_019763.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032157.mm.1 |
TC0400003884.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032157.mm.1 |
NM_019763.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032157.mm.1 |
NM_019763.2 |
Refseq |