Assay Details
| Gene Symbol: Spen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019763.2 | NP_062737.2 | 4 - 5 | 1394 | 86 | |
| GenBank mRNA | AB055980.1 | - | 3 - 4 | 836 | 86 | |
| AK137647.1 | - | 4 - 5 | 1282 | 86 | ||
| Z78160.1 | - | 1 - 2 | 90 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spen
Genomic Map