Assay Details
| Gene Symbol: Spata21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177867.3 | NP_808535.1 | 9 - 10 | 1660 | 65 | |
| GenBank mRNA | AK077155.1 | - | 9 - 10 | 1661 | 65 | |
| BC112427.1 | - | 8 - 9 | 1369 | 40062127 | 65 | |
| BC113132.1 | - | 8 - 9 | 1369 | 40062125 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spata21
Genomic Map