| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400014778.mm.1 |
NM_172520.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014778.mm.1 |
NM_172520.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014778.mm.1 |
TC0400001783.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014777.mm.1 |
NM_172520.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014777.mm.1 |
NM_172520.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014777.mm.1 |
TC0400001783.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014775.mm.1 |
NM_172520.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014775.mm.1 |
NM_172520.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014775.mm.1 |
TC0400001783.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014773.mm.1 |
NM_172520.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014773.mm.1 |
NM_172520.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014773.mm.1 |
TC0400001783.mm.1 |
MTA TC |