Assay Details
| Gene Symbol: Arhgef19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172520.2 | NP_766108.1 | 6 - 7 | 1180 | 56 | |
| GenBank mRNA | AK031443.1 | - | 6 - 7 | 1181 | 56 | |
| AK053075.1 | - | 6 - 7 | 1181 | 56 | ||
| BC060376.1 | - | 4 - 5 | 910 | 5340007 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef19
Genomic Map