Assay Details
| Gene Symbol: Atf7ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153123.2 | NP_694763.2 | 10 - 11 | 1077 | 133 | |
| GenBank mRNA | AK145634.1 | - | 10 - 11 | 1036 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atf7ip2
Genomic Map