Assay Details
| Gene Symbol: Atf7ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029253.1 | NP_083529.1 | 1 - 2 | 117 | 132 | |
| NM_153123.2 | NP_694763.2 | 1 - 2 | 117 | 132 | ||
| GenBank mRNA | AK016180.1 | - | 1 - 2 | 119 | 132 | |
| AK049401.1 | - | 1 - 2 | 76 | 132 | ||
| AK077229.1 | - | 1 - 2 | 77 | 132 | ||
| AK139862.1 | - | 1 - 2 | 74 | 132 | ||
| AK145634.1 | - | 1 - 2 | 76 | 132 | ||
| BY715784.1 | - | 1 - 2 | 119 | 132 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016180.1, AK077229.1, AK139862.1, AK145634.1, NM_029253.1, NM_153123.2 transcripts.
Set Membership: |
|

Atf7ip2
Genomic Map