Assay Details
| Gene Symbol: Atf7ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029253.1 | NP_083529.1 | 3 - 4 | 392 | 65 | |
| NM_153123.2 | NP_694763.2 | 3 - 4 | 392 | 65 | ||
| GenBank mRNA | AK016180.1 | - | 3 - 4 | 394 | 65 | |
| AK049401.1 | - | 3 - 4 | 351 | 65 | ||
| AK077229.1 | - | 3 - 4 | 352 | 65 | ||
| AK139862.1 | - | 3 - 4 | 349 | 65 | ||
| AK145634.1 | - | 3 - 4 | 351 | 65 | ||
| BY715784.1 | - | 3 - 4 | 394 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378511 | BC018510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378511 | AK145634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378511 | NM_153123.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378511 | BC018510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378511 | AK145634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378511 | NM_153123.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005893 | AK145634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005893 | AY903215.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005893 | AK077229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005893 | NM_153123.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005893 | AK016180.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763487 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633816 | BC076625.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633816 | BC085176.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633816 | DQ192038.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633816 | NM_001007580.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114842 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180512 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168884 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177887 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005893 | AK145634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005893 | AY903215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005893 | AK077229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005893 | NM_153123.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005893 | AK016180.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378511 | BC018510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378511 | AK145634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378511 | NM_153123.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378511 | BC018510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378511 | AK145634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378511 | NM_153123.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08505940 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177887 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168884 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08505917 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08505962 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180512 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08505909 | - | - |
| Whole Mouse Genome G4122A | A_51_P196148 | AY903215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196148 | AK077229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P620766 | BY715784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P620766 | AK139862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P620766 | AK145634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P620766 | AK077229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P620766 | NM_153123.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P247233 | AK049401.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168884 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177887 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180512 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08505940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12651199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12920797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12656454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12920796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12597686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12538590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30541072 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433489 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433493 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427603_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430364_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442796_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433492 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433484 | - | - |
| Mouse Genome 430 2.0 Array | 1430364_at | - | - |
| Mouse Genome 430 2.0 Array | 1427603_at | - | - |
| Mouse Genome 430 2.0 Array | 1427603_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442796_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430364_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442796_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75329_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75329_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600000754.mm.1 | NM_029253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000754.mm.1 | NM_029253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000754.mm.1 | NM_153123.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000754.mm.1 | NM_153123.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000754.mm.1 | TC1600000110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600000753.mm.1 | NM_029253.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000753.mm.1 | NM_029253.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000753.mm.1 | NM_153123.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000753.mm.1 | NM_153123.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000753.mm.1 | TC1600000110.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1442796_PM_at | - | - |
| Mouse Expression Set 430 | 1430364_PM_at | - | - |
| Mouse Expression Set 430 | 1442796_at | - | - |
| Mouse Expression Set 430 | 1430364_at | - | - |
| Murine Genome U74v2 Set | 168286_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168286_i_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427603_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427603_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442796_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430364_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442796_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430364_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 534488 | - | - |
| ABI Mouse Genome Survey Microarray | 696550 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3290215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107050390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687679 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620418 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2620418 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2687679 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229033 | - | - |
| mouse-6 v1.1 (Illumina) | scl0223963.1_197-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290215 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107050390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0223963.1_197-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107050390 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2620418 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2620418 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229033 | - | - |

Atf7ip2
Genomic Map