| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600001063.mm.1 |
NM_001122667.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001063.mm.1 |
NM_001122667.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001063.mm.1 |
NM_181860.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001063.mm.1 |
NM_181860.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001063.mm.1 |
TC1600000150.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600001060.mm.1 |
NM_001122667.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001060.mm.1 |
NM_001122667.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001060.mm.1 |
TC1600000150.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600001059.mm.1 |
NM_001122667.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001059.mm.1 |
NM_001122667.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001059.mm.1 |
TC1600000150.mm.1 |
MTA TC |