Assay Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122667.2 | NP_001116139.1 | 2 - 3 | 132 | 78 | |
| NM_181860.1 | NP_862908.1 | 1 - 2 | 68 | 78 | ||
| GenBank mRNA | BC028459.1 | - | 1 - 2 | 68 | 78 | |
| X66901.1 | - | 2 - 3 | 115 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC028459.1, NM_001122667.2, NM_181860.1, X66901.1 transcripts.
Set Membership: |
|

Mkl2
Genomic Map