Assay Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122667.2 | NP_001116139.1 | 7 - 8 | 687 | 69 | |
| NM_153588.3 | NP_705816.2 | 5 - 6 | 701 | 69 | ||
| GenBank mRNA | AF532598.1 | - | 5 - 6 | 483 | 69 | |
| AK220377.1 | - | 5 - 6 | 483 | 69 | ||
| BC131644.1 | - | 5 - 6 | 483 | 40110019 | 69 | |
| BC131645.1 | - | 5 - 6 | 481 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkl2
Genomic Map