Assay Details
| Gene Symbol: Ddx41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134059.2 | NP_598820.2 | 13 - 14 | 1552 | 65 | |
| GenBank mRNA | AK151427.1 | - | 13 - 14 | 1426 | 65 | |
| AK156957.1 | - | 13 - 14 | 1420 | 65 | ||
| AK168043.1 | - | 13 - 14 | 1425 | 65 | ||
| BC011308.1 | - | 13 - 14 | 1406 | 3256762 | 65 | |
| BC023711.1 | - | 13 - 14 | 1427 | 65 | ||
| BE864921.1 | - | 4 - 5 | 343 | 65 | ||
| CX231996.1 | - | 6 - 7 | 745 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx41
Genomic Map