Assay Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142152.1 | - | 11 - 12 | 1333 | 73 | |
| AK154645.1 | - | 8 - 9 | 3573 | 73 | ||
| BC031484.1 | - | 11 - 12 | 1331 | 2654265 | 73 | |
| BC046438.1 | - | 3 - 4 | 307 | 73 | ||
| BC047372.1 | - | 11 - 12 | 1314 | 73 | ||
| BC057597.1 | - | 9 - 10 | 2899 | 73 | ||
| BC089379.1 | - | 11 - 12 | 1288 | 73 | ||
More Information
Set Membership: |
|

Brd9
Genomic Map