Assay Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142152.1 | - | 1 - 2 | 112 | 58 | |
| AK154645.1 | - | 1 - 2 | 126 | 58 | ||
| BC031484.1 | - | 1 - 2 | 107 | 2654265 | 58 | |
| BC047372.1 | - | 1 - 2 | 91 | 58 | ||
| BC057597.1 | - | 1 - 2 | 88 | 58 | ||
| BC089379.1 | - | 1 - 2 | 65 | 58 | ||
| BY035489.1 | - | 1 - 2 | 136 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brd9
Genomic Map