Assay Details
| Gene Symbol: Reck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016678.2 | NP_057887.2 | 3 - 4 | 312 | 62 | |
| GenBank mRNA | AB006960.1 | - | 3 - 4 | 242 | 62 | |
| AK042266.1 | - | 3 - 4 | 313 | 62 | ||
| AK143657.1 | - | 3 - 4 | 290 | 62 | ||
| AK157885.1 | - | 3 - 4 | 300 | 62 | ||
| BC053025.1 | - | 3 - 4 | 251 | 62 | ||
| BC132197.1 | - | 3 - 4 | 340 | 40130474 | 62 | |
| BC138065.1 | - | 3 - 4 | 340 | 8861084 | 62 | |
| BC145133.1 | - | 3 - 4 | 340 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Reck
Genomic Map