Assay Details
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013739.2 | NP_038767.1 | 4 - 5 | 675 | 59 | |
GenBank mRNA | AF179242.1 | - | 4 - 5 | 675 | 59 | |
AF237580.1 | - | 4 - 5 | 671 | 59 | ||
AK137520.1 | - | 4 - 5 | 662 | 59 | ||
AK154750.1 | - | 3 - 4 | 1498 | 59 | ||
AK155180.1 | - | 3 - 4 | 1506 | 59 | ||
AK156060.1 | - | 3 - 4 | 1505 | 59 | ||
AK172511.1 | - | 3 - 4 | 1505 | 59 | ||
BC048784.1 | - | 3 - 4 | 1806 | 59 | ||
BC116922.1 | - | 4 - 5 | 674 | 40126241 | 59 | |
BC120535.1 | - | 4 - 5 | 674 | 40129458 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | BC048784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | BC116922.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | BC120535.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | AK156060.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | AK172511.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | BC048784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | BC116922.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | BC120535.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | AK156060.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P578790 | AK172511.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30491 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492608 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607851 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129754 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171069 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | BC048784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | BC116922.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | BC120535.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | AK156060.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | AK172511.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | BC048784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | BC116922.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | BC120535.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | AK156060.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P578790 | AK172511.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055577 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055581 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055588 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055579 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29055598 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171069 | - | - |
Whole Mouse Genome G4122A | A_52_P578790 | BC048784.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | BC116922.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | BC120535.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | AK156060.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | AK172511.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | BC048784.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | BC116922.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | BC120535.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | AK156060.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P578790 | AK172511.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100128 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171069 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10973723 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10704573 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10704572 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10973733 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10973728 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10973718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29055565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10973725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10973737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10797120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10774722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055582 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055561 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10797115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10973719 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10973729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10710300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10710292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29055596 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10409503 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409507 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409505 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409504 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409506 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418096_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10409502 | - | - |
Mouse Genome 430 2.0 Array | 1418096_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418096_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27261_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27261_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300013059.mm.1 | NM_013739.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300013059.mm.1 | NM_013739.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300013059.mm.1 | TC1300002062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300013058.mm.1 | NM_013739.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300013058.mm.1 | NM_013739.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300013058.mm.1 | TC1300002062.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418096_at | - | - |
Mouse Expression Set 430 | 1418096_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418096_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418096_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418096_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418096_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 130347 | - | - |
MouseWG-6 v2 Expression BeadChip | 3870167 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3870167 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 130347 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003870167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000130347 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258357 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2681443 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258357 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2681443 | - | - |
mouse-6 v1.1 (Illumina) | scl0003686.1_14-S | - | - |
mouse-6 v1.1 (Illumina) | scl43920.5.585_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3870167 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 130347 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003686.1_14-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl43920.5.585_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870167 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130347 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1258357 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258357 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl43920.5.585_30-S | - | - |
