Assay Details
Gene Symbol: Caml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007596.2 | NP_031622.2 | 2 - 3 | 719 | 107 | |
GenBank mRNA | AK002895.1 | - | 2 - 3 | 717 | 107 | |
BC005688.1 | - | 2 - 3 | 642 | 3156307 | 107 | |
U21960.1 | - | 2 - 3 | 667 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
