Assay Details
| Gene Symbol: Efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025769.3 | NP_080045.1 | 2 - 3 | 331 | 142 | |
| GenBank mRNA | AK015426.1 | - | 313 | 142 | ||
| AK015866.1 | - | 2 - 3 | 351 | 142 | ||
| AK017267.1 | - | 2 - 3 | 332 | 142 | ||
| AK030381.1 | - | 2 - 3 | 341 | 142 | ||
| AK038735.1 | - | 2 - 3 | 341 | 142 | ||
| AK042748.1 | - | 2 - 3 | 373 | 142 | ||
| AK043519.1 | - | 2 - 3 | 303 | 142 | ||
| AK077722.1 | - | 2 - 3 | 334 | 142 | ||
| BC085167.1 | - | 2 - 3 | 223 | 30538702 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab1
Genomic Map