Assay Details
| Gene Symbol: P3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173379.2 | NP_775555.1 | 3 - 4 | 906 | 117 | |
| GenBank mRNA | AJ430350.1 | - | 3 - 4 | 907 | 117 | |
More Information
Set Membership: |
|

P3h2
Genomic Map