Assay Details
| Gene Symbol: P3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173379.2 | NP_775555.1 | 5 - 6 | 1186 | 97 | |
| GenBank mRNA | AJ430350.1 | - | 5 - 6 | 1187 | 97 | |
| AK076434.1 | - | 204 | 97 | |||
More Information
Updated Mapping Information
Set Membership: |
|

P3h2
Genomic Map