Assay Details
| Gene Symbol: Tmem207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101640.1 | NP_001095110.1 | 1 - 2 | 135 | 98 | |
| GenBank mRNA | AK142524.1 | - | 1 - 2 | 134 | 98 | |
More Information
Set Membership: |
|

Tmem207
Genomic Map