Assay Details
| Gene Symbol: Rasal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178785.3 | NP_848900.2 | 14 - 15 | 2877 | 122 | |
| GenBank mRNA | AK030797.1 | - | 14 - 15 | 2880 | 122 | |
| AK088449.1 | - | 14 - 15 | 2839 | 122 | ||
| AK088987.1 | - | 14 - 15 | 2855 | 122 | ||
| AK131128.1 | - | 16 - 17 | 2784 | 122 | ||
| BC132341.1 | - | 16 - 17 | 2783 | 40130618 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasal3
Genomic Map