Assay Details
| Gene Symbol: Rasal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178785.3 | NP_848900.2 | 8 - 9 | 1638 | 98 | |
| GenBank mRNA | AK030797.1 | - | 8 - 9 | 1641 | 98 | |
| AK040573.1 | - | 8 - 9 | 1626 | 98 | ||
| AK041479.1 | - | 9 - 10 | 1496 | 98 | ||
| AK088449.1 | - | 8 - 9 | 1600 | 98 | ||
| AK088987.1 | - | 8 - 9 | 1616 | 98 | ||
| AK131128.1 | - | 10 - 11 | 1545 | 98 | ||
| BC132341.1 | - | 10 - 11 | 1544 | 40130618 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasal3
Genomic Map