Assay Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010061.5 | NP_034191.3 | 4 - 5 | 611 | 92 | |
| GenBank mRNA | AK157889.1 | - | 535 | 92 | ||
| BC030394.1 | - | 4 - 5 | 628 | 92 | ||
| BC115570.1 | - | 4 - 5 | 400 | 40102105 | 92 | |
| BC115571.1 | - | 4 - 5 | 400 | 40102106 | 92 | |
| D83038.1 | - | 5 - 6 | 481 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1
Genomic Map