Assay Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010061.5 | NP_034191.3 | 5 - 6 | 734 | 69 | |
| GenBank mRNA | AK157889.1 | - | 658 | 69 | ||
| BC014718.1 | - | 5 - 6 | 725 | 4925690 | 69 | |
| BC030394.1 | - | 5 - 6 | 751 | 69 | ||
| BC115570.1 | - | 5 - 6 | 523 | 40102105 | 69 | |
| BC115571.1 | - | 5 - 6 | 523 | 40102106 | 69 | |
| D83038.1 | - | 6 - 7 | 604 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1
Genomic Map