Assay Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010061.5 | NP_034191.3 | 6 - 7 | 841 | 79 | |
| GenBank mRNA | AK157889.1 | - | 765 | 79 | ||
| BC014718.1 | - | 6 - 7 | 832 | 4925690 | 79 | |
| BC030394.1 | - | 6 - 7 | 858 | 79 | ||
| BC115570.1 | - | 6 - 7 | 630 | 40102105 | 79 | |
| BC115571.1 | - | 6 - 7 | 630 | 40102106 | 79 | |
| D83038.1 | - | 7 - 8 | 711 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA catabolic process, endonucleolytic
DNA metabolic process
DNA catabolic process
apoptotic process
nucleic acid phosphodiester bond hydrolysis
actin binding
nuclease activity
endonuclease activity
endodeoxyribonuclease activity
deoxyribonuclease I activity
deoxyribonuclease activity
hydrolase activity
endodeoxyribonuclease activity, producing 5'-phosphomonoesters
DNA catabolic process, endonucleolytic
DNA metabolic process
DNA catabolic process
apoptotic process
nucleic acid phosphodiester bond hydrolysis
actin binding
nuclease activity
endonuclease activity
endodeoxyribonuclease activity
deoxyribonuclease I activity
deoxyribonuclease activity
hydrolase activity
endodeoxyribonuclease activity, producing 5'-phosphomonoesters
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985835 | BC030394.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985835 | BX638483.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985835 | NM_010061.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289702 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133499 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985835 | BC030394.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985835 | BX638483.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985835 | NM_010061.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358445 | U00478.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358445 | BC030394.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358445 | BC115570.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358445 | BC115571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P358445 | BX638483.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30507829 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133499 | - | - |
| Whole Mouse Genome G4122A | A_51_P358445 | U00478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358445 | BC030394.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358445 | BC115570.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358445 | BC115571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358445 | BX638483.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127357 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133499 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13003312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13003318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13003333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13003325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13003284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13003307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30507798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12614527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12974781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12713532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13003299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12523334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12997891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12638023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13003320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12974780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13003313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13003284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13003285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12974783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13003330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30507862 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433248 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424592_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433241 | - | - |
| Mouse Genome 430 2.0 Array | 1424592_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424592_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13419_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13419_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa1400_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600000181.mm.1 | NM_010061.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000181.mm.1 | NM_010061.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000181.mm.1 | TC1600000022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600000178.mm.1 | NM_010061.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000178.mm.1 | NM_010061.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000178.mm.1 | TC1600000022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600000180.mm.1 | NM_010061.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000180.mm.1 | NM_010061.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000180.mm.1 | TC1600000022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600000175.mm.1 | NM_010061.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000175.mm.1 | NM_010061.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000175.mm.1 | TC1600000022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600000182.mm.1 | NM_010061.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000182.mm.1 | NM_010061.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000182.mm.1 | TC1600000022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600000174.mm.1 | NM_010061.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000174.mm.1 | NM_010061.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600000174.mm.1 | TC1600000022.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424592_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424592_a_at | - | - |
| Murine Genome U74v2 Set | 92865_PM_at | - | - |
| Murine Genome U74v2 Set | 92865_at | - | - |
| Murine Genome U74v2 Set | 160452_PM_at | - | - |
| Murine Genome U74v2 Set | 160452_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424592_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424592_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424592_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424592_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6840546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1190167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190167 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649560 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734251 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734251 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649560 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001775.1_4-S | - | - |
| mouse-6 v1.1 (Illumina) | scl49480.1_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190167 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001775.1_4-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49480.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840546 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734251 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734251 | - | - |

Dnase1
Genomic Map