Assay Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293648.1 | NP_001280577.1 | 11 - 12 | 1883 | 86 | |
| NM_019676.3 | NP_062650.1 | 11 - 12 | 1834 | 86 | ||
| NR_121571.1 | 11 - 12 | 1892 | 86 | |||
| GenBank mRNA | AF133125.1 | - | 11 - 12 | 1816 | 86 | |
| AK028749.1 | - | 11 - 12 | 1835 | 86 | ||
| AK082890.1 | - | 11 - 12 | 1826 | 86 | ||
| BC015249.1 | - | 11 - 12 | 1846 | 4241580 | 86 | |
| BC025798.1 | - | 11 - 12 | 1810 | 5068610 | 86 | |
| JN635596.1 | - | 11 - 12 | 1883 | 86 | ||
| U85711.1 | - | 11 - 12 | 1828 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plcd1
Genomic Map