Assay Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293648.1 | NP_001280577.1 | 3 - 4 | 591 | 111 | |
| NM_019676.3 | NP_062650.1 | 3 - 4 | 542 | 111 | ||
| NR_121571.1 | 3 - 4 | 542 | 111 | |||
| GenBank mRNA | AF133125.1 | - | 3 - 4 | 524 | 111 | |
| AK028749.1 | - | 3 - 4 | 543 | 111 | ||
| AK082890.1 | - | 3 - 4 | 534 | 111 | ||
| BC015249.1 | - | 3 - 4 | 496 | 4241580 | 111 | |
| BC025798.1 | - | 3 - 4 | 518 | 5068610 | 111 | |
| JN635596.1 | - | 3 - 4 | 591 | 111 | ||
| U85711.1 | - | 3 - 4 | 536 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plcd1
Genomic Map