Assay Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011049.4 | NP_035179.1 | 6 - 7 | 957 | 57 | |
| GenBank mRNA | AK051425.1 | - | 6 - 7 | 958 | 57 | |
| AK145483.1 | - | 6 - 7 | 987 | 57 | ||
| AK166188.1 | - | 6 - 7 | 1166 | 57 | ||
| AK214881.1 | - | 1 - 2 | 73 | 57 | ||
| BC011069.1 | - | 6 - 7 | 700 | 4006745 | 57 | |
| BC013663.1 | - | 6 - 7 | 1016 | 4208974 | 57 | |
| X69025.1 | - | 6 - 7 | 979 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk16
Genomic Map