Assay Details
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290643.1 | NP_001277572.1 | 1 - 2 | 350 | 96 | |
| NM_133669.5 | NP_598430.1 | 1 - 2 | 350 | 96 | ||
| NR_110966.1 | 1 - 2 | 350 | 96 | |||
| NR_110967.1 | 1 - 2 | 350 | 96 | |||
| GenBank mRNA | AJ303371.1 | - | 1 - 2 | 176 | 96 | |
| AK017422.1 | - | 1 - 2 | 218 | 96 | ||
| AK044002.1 | - | 1 - 2 | 257 | 96 | ||
| AK050418.1 | - | 1 - 2 | 224 | 96 | ||
| AK076907.1 | - | 1 - 2 | 212 | 96 | ||
| AK132713.1 | - | 1 - 2 | 350 | 96 | ||
| AK162309.1 | - | 1 - 2 | 268 | 96 | ||
| BC049698.1 | - | 1 - 2 | 228 | 6706696 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rp2h
Genomic Map