Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 11 - 12 | 1517 | 95 | |
| GenBank mRNA | AK031767.1 | - | 11 - 12 | 1496 | 95 | |
| AK049053.1 | - | 11 - 12 | 1931 | 95 | ||
| AK169116.1 | - | 3 - 4 | 340 | 95 | ||
| AK220478.1 | - | 10 - 11 | 1230 | 95 | ||
| BC005470.1 | - | 9 - 10 | 1086 | 3496607 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map