Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 16 - 17 | 2222 | 73 | |
| GenBank mRNA | AK031767.1 | - | 16 - 17 | 2201 | 73 | |
| AK169116.1 | - | 8 - 9 | 1045 | 73 | ||
| AK172230.1 | - | 3 - 4 | 1185 | 73 | ||
| AK220478.1 | - | 14 - 15 | 2737 | 73 | ||
| BC005470.1 | - | 14 - 15 | 1791 | 3496607 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map