Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 18 - 19 | 2486 | 138 | |
| GenBank mRNA | AK031767.1 | - | 18 - 19 | 2465 | 138 | |
| AK169116.1 | - | 10 - 11 | 1309 | 138 | ||
| AK172230.1 | - | 5 - 6 | 1449 | 138 | ||
| AK220478.1 | - | 16 - 17 | 3001 | 138 | ||
| BC005470.1 | - | 16 - 17 | 2055 | 3496607 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map