Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 19 - 20 | 2601 | 83 | |
| GenBank mRNA | AK031767.1 | - | 19 - 20 | 2580 | 83 | |
| AK169116.1 | - | 11 - 12 | 1424 | 83 | ||
| AK172230.1 | - | 6 - 7 | 1564 | 83 | ||
| AK220478.1 | - | 17 - 18 | 3116 | 83 | ||
| BC005470.1 | - | 17 - 18 | 2170 | 3496607 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map