Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 2 - 3 | 322 | 90 | |
| GenBank mRNA | AK031767.1 | - | 2 - 3 | 300 | 90 | |
| AK049053.1 | - | 3 - 4 | 343 | 90 | ||
| AK166523.1 | - | 2 - 3 | 298 | 90 | ||
| AK220478.1 | - | 1 - 2 | 35 | 90 | ||
| BY272535.1 | - | 2 - 3 | 323 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031767.1, AK049053.1 transcripts.
Set Membership: |
|

Usp11
Genomic Map