Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 8 - 9 | 1058 | 85 | |
| GenBank mRNA | AK031767.1 | - | 8 - 9 | 1037 | 85 | |
| AK049053.1 | - | 8 - 9 | 1472 | 85 | ||
| AK166523.1 | - | 8 - 9 | 1034 | 85 | ||
| AK220478.1 | - | 7 - 8 | 771 | 85 | ||
| BC005470.1 | - | 6 - 7 | 627 | 3496607 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map