Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 9 - 10 | 1215 | 137 | |
| GenBank mRNA | AK031767.1 | - | 9 - 10 | 1194 | 137 | |
| AK049053.1 | - | 9 - 10 | 1629 | 137 | ||
| AK220478.1 | - | 8 - 9 | 928 | 137 | ||
| BC005470.1 | - | 7 - 8 | 784 | 3496607 | 137 | |
More Information
Set Membership: |
|

Usp11
Genomic Map