Assay Details
| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134189.2 | NP_598950.2 | 5 - 6 | 873 | 76 | |
| GenBank mRNA | AK033515.1 | - | 4 - 5 | 531 | 76 | |
| AK131155.1 | - | 5 - 6 | 851 | 76 | ||
| AK143714.1 | - | 4 - 5 | 704 | 76 | ||
| AK159088.1 | - | 5 - 6 | 872 | 76 | ||
| AK159362.1 | - | 5 - 6 | 871 | 76 | ||
| AK162424.1 | - | 5 - 6 | 873 | 76 | ||
| BC060617.1 | - | 5 - 6 | 856 | 6836791 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt10
Genomic Map