Assay Details
| Gene Symbol: Sap30l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081168.1 | NP_001074637.1 | 3 - 4 | 876 | 75 | |
| GenBank mRNA | AK010234.1 | - | 3 - 4 | 364 | 75 | |
| AK203234.1 | - | 1 - 2 | 64 | 75 | ||
| BC051686.1 | - | 3 - 4 | 876 | 6433799 | 75 | |
| BC094930.1 | - | 4 - 5 | 595 | 75 | ||
| BC117029.1 | - | 3 - 4 | 590 | 40126348 | 75 | |
| BC117031.1 | - | 3 - 4 | 590 | 40126350 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827570 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827569 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827571 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827566 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | NM_001081168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | AK010234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | BC051686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | BC094930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | NM_001081168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | AK010234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | BC051686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241943 | BC094930.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91007 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151781 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | NM_001081168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | AK010234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | BC051686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | BC094930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | NM_001081168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | AK010234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | BC051686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241943 | BC094930.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151781 | - | - |
| Whole Mouse Genome G4122A | A_51_P241943 | NM_001081168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | AK010234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | BC051686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | BC094930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | NM_001081168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | AK010234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | BC051686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241943 | BC094930.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151781 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827570 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827571 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27827569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27827638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09425043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09003657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08989364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09000205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09003647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09136180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09181691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09089692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09086049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09234780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09743273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09438827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09367394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09136187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09142680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08989355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09181696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09161495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09743276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09438820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09675017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09484352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09094281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09142684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09410714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09086051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27827686 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10376289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376288 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376285 | - | - |
| Mouse Genome 430 2.0 Array | 1447379_at | - | - |
| Mouse Genome 430 2.0 Array | 1415718_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447379_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415718_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50724_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50724_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100005365.mm.1 | NM_001081168.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005365.mm.1 | NM_001081168.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005365.mm.1 | TC1100000668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100005367.mm.1 | NM_001081168.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005367.mm.1 | NM_001081168.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005367.mm.1 | TC1100000668.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100005363.mm.1 | NM_001081168.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005363.mm.1 | NM_001081168.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005363.mm.1 | TC1100000668.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415718_at | - | - |
| Mouse Expression Set 430 | 1415718_PM_at | - | - |
| Mouse Expression Set 430 | 1415718_PM_at | - | - |
| Mouse Expression Set 430 | 1415718_at | - | - |
| Murine Genome U74v2 Set | 104453_PM_at | - | - |
| Murine Genome U74v2 Set | 104453_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415718_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415718_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 616336 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2759789 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2759789 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3128784 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3128784 | - | - |

Sap30l
Genomic Map