Assay Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011672.4 | NP_035802.3 | 9 - 10 | 794 | 73 | |
| NR_028403.1 | 9 - 10 | 768 | 73 | |||
| GenBank mRNA | AK010461.1 | - | 9 - 10 | 763 | 73 | |
| AK012543.1 | - | 7 - 8 | 738 | 73 | ||
| AK088823.1 | - | 9 - 10 | 763 | 73 | ||
| AK147138.1 | - | 9 - 10 | 777 | 73 | ||
| AK169062.1 | - | 9 - 10 | 776 | 73 | ||
| BC006630.1 | - | 9 - 10 | 751 | 3708320 | 73 | |
| U64445.1 | - | 9 - 10 | 73 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ufd1l
Genomic Map