Assay Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011672.4 | NP_035802.3 | 11 - 12 | 970 | 87 | |
| NR_028403.1 | 11 - 12 | 944 | 87 | |||
| GenBank mRNA | AK010461.1 | - | 11 - 12 | 939 | 87 | |
| AK012543.1 | - | 9 - 10 | 914 | 87 | ||
| AK088823.1 | - | 11 - 12 | 939 | 87 | ||
| AK147138.1 | - | 11 - 12 | 953 | 87 | ||
| AK169062.1 | - | 11 - 12 | 952 | 87 | ||
| BC006630.1 | - | 11 - 12 | 927 | 3708320 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | U64445.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | AK088823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | AK147138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | NM_011672.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | NR_028403.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | U64445.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | AK088823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | AK147138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | NM_011672.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P220347 | NR_028403.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047509 | BC006630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047509 | AK012543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047509 | AK088823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047509 | AK147138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047509 | AK169062.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160531 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155387 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103079 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047509 | BC006630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047509 | AK012543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047509 | AK088823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047509 | AK147138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047509 | AK169062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | U64445.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | AK088823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | AK147138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | NM_011672.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | NR_028403.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | U64445.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | AK088823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | AK147138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | NM_011672.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P220347 | NR_028403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | U64445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | AK088823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | AK147138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | NM_011672.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | NR_028403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | U64445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | AK088823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | AK147138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | NM_011672.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P220347 | NR_028403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P328963 | U64445.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13006414 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13006429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13006415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13006402 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10434237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434236 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434247 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434240 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418087_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432367_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10434233 | - | - |
| Mouse Genome 430 2.0 Array | 1418087_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418087_at | - | - |
| Mouse Genome 430 2.0 Array | 1432367_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432367_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22230_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22230_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600002323.mm.1 | NM_011672.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002323.mm.1 | NM_011672.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002323.mm.1 | NR_028403.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002323.mm.1 | NR_028403.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002323.mm.1 | TC1600000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600002322.mm.1 | NM_011672.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002322.mm.1 | NM_011672.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002322.mm.1 | NR_028403.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002322.mm.1 | NR_028403.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002322.mm.1 | TC1600000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600002325.mm.1 | NM_011672.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002325.mm.1 | NM_011672.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002325.mm.1 | NR_028403.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002325.mm.1 | NR_028403.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002325.mm.1 | TC1600000250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600002324.mm.1 | NM_011672.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002324.mm.1 | NM_011672.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002324.mm.1 | NR_028403.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002324.mm.1 | NR_028403.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002324.mm.1 | TC1600000250.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418087_PM_at | - | - |
| Mouse Expression Set 430 | 1432367_PM_a_at | - | - |
| Mouse Expression Set 430 | 1418087_at | - | - |
| Mouse Expression Set 430 | 1432367_a_at | - | - |
| Murine Genome U74v2 Set | 93303_at | - | - |
| Murine Genome U74v2 Set | 93303_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432367_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432367_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418087_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418087_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418087_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432367_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432367_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418087_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2443482 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2443482 | - | - |

Ufd1l
Genomic Map