Assay Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011672.4 | NP_035802.3 | 6 - 7 | 619 | 76 | |
| NR_028403.1 | 6 - 7 | 593 | 76 | |||
| GenBank mRNA | AK010461.1 | - | 6 - 7 | 588 | 76 | |
| AK012543.1 | - | 4 - 5 | 563 | 76 | ||
| AK088823.1 | - | 6 - 7 | 588 | 76 | ||
| AK147138.1 | - | 6 - 7 | 602 | 76 | ||
| AK169062.1 | - | 6 - 7 | 601 | 76 | ||
| BC006630.1 | - | 6 - 7 | 576 | 3708320 | 76 | |
More Information
Set Membership: |
|

Ufd1l
Genomic Map