Assay Details
| Gene Symbol: Tbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285472.1 | NP_001272401.1 | 2 - 3 | 163 | 66 | |
| GenBank mRNA | AF349658.1 | - | 2 - 3 | 163 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF349658.1, NM_001285472.1 transcripts.
Set Membership: |
|

Tbx1
Genomic Map